Learn More
ATCC Quantitative Genomic DNA from; Campylobacter jejuni; subsp. jejuni; Strain: AS-83-79
Catalog No. 502384384
Encompass
Quantitative genomic DNA from Campylobacter jejuni subsp. jejuni can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4384 Supplier ATCC Supplier No. 33291DQ
Quantitative genomic DNA from Campylobacter jejuni subsp. jejuni can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.