Learn More
ATCC Quantitative Genomic DNA from; Clostridium sporogenes; Strain: L.S. McClung 2006
Catalog No. 502384806
Encompass
Quantitative Genomic DNA from Clostridium sporogenes can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4806 Supplier ATCC Supplier No. 11437DQ
Quantitative Genomic DNA from Clostridium sporogenes can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.