Learn More
ATCC Quantitative Genomic DNA from; Saccharomyces cerevisiae; Strain: EBY100
Supplier:  ATCC MYA4941DQ
Encompass
Quantitative Genomic DNA from Saccharomyces cerevisiae strain EBY100 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.
Catalog No. 50-238-4896