Learn More
ATCC Staphylococcus simulans
Catalog No. 502380894
Encompass
Staphylococcus simulans strain MK 148 is a bacterial type strain that was isolated from human skin. This strain is propagated aerobically.
Catalog No. 50-238-0894 Supplier ATCC Supplier No. 27848
Staphylococcus simulans strain MK 148 is a bacterial type strain that was isolated from human skin. This strain is propagated aerobically.