Learn More
ATCC Merck Pneumococcal polysaccharide powder Type 9 (d9N) 2mg
Catalog No. 502380612
Encompass
Purified pneumococcal polysaccharides are manufactured by Merck & Company for small quantity distribution by ATCC. The pneumococcal powders include those types which are components of the current 23-valent vaccine against Streptococcus pneumoniae.Each of the polysaccharide antigens used in the polyvalent vaccine is extracted and purified separately. These vaccine products can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.
Catalog No. 50-238-0612 Supplier ATCC Supplier No. 21X
Purified pneumococcal polysaccharides are manufactured by Merck & Company for small quantity distribution by ATCC. The pneumococcal powders include those types which are components of the current 23-valent vaccine against Streptococcus pneumoniae.Each of the polysaccharide antigens used in the polyvalent vaccine is extracted and purified separately. These vaccine products can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.