Learn More
ATCC Ruminococcus callidus
Catalog No. 502381192
Encompass
Ruminococcus callidus strain VPI S7-31 is a bacterial type strain that was isolated from human feces. This strain is propagated anaerobically and grows best when meat particles are left in the broth and sterile rumen fluid is added at the time of inoculation
Catalog No. 50-238-1192 Supplier ATCC Supplier No. 27760
Ruminococcus callidus strain VPI S7-31 is a bacterial type strain that was isolated from human feces. This strain is propagated anaerobically and grows best when meat particles are left in the broth and sterile rumen fluid is added at the time of inoculation