Learn More
ATCC Quantitative Genomic DNA; from Babesia microti; Strain: GI (Ingram strain)
Catalog No. 502382438
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load.
Catalog No. 50-238-2438 Supplier ATCC Supplier No. PRA398DQ
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load.