Learn More
ATCC Quantitative Genomic; DNA from; Chlamydia trachomatis; Serovar D strain UW-3/Cx
Catalog No. 502381863
Encompass
Genomic DNA isolated from a preparation of Chlamydia trachomatis Serovar D strain UW-3/Cx (ATCC VR-885). This product can be used for assay development, verification and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-1863 Supplier ATCC Supplier No. VR885DQ
Genomic DNA isolated from a preparation of Chlamydia trachomatis Serovar D strain UW-3/Cx (ATCC VR-885). This product can be used for assay development, verification and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.