Learn More
ATCC Quantitative; Genomic DNA; from BK Polyomavirus
Supplier:  ATCC VR837DQ
Encompass
Quantitative Genomic DNA from BK Polyomavirus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4042