Learn More
ATCC Quantitative Genomic DNA from; Gardnerella vaginalis; Strain AmMS 117
Catalog No. 502382433
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacteria load.
Catalog No. 50-238-2433 Supplier ATCC Supplier No. 49145DQ
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacteria load.