Learn More
ATCC Quantitative Genomic; DNA from Human; adenovirus 5; Strain: Adenoid 75
Catalog No. 502385106
Encompass
Quantitative genomic DNA from Human adenovirus 5 strain Adenoid 75 can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-5106 Supplier ATCC Supplier No. VR5DQ
Quantitative genomic DNA from Human adenovirus 5 strain Adenoid 75 can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.