Learn More
ATCC Quantitative Genomic; RNA from Human; parainfluenza virus 2; Strain Greer
Catalog No. 502383022
Encompass
Quantitative Genomic RNA from Human parainfluenza virus 2 strain Greer can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3022 Supplier ATCC Supplier No. VR92DQ
Quantitative Genomic RNA from Human parainfluenza virus 2 strain Greer can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.