Learn More
ATCC Quantitative Genomic; RNA from Human; respiratory syncytial; virus strain 9320
Catalog No. 502382124
Encompass
Quantitative Genomic RNA from Human respiratory syncytial virus strain 9320 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2124 Supplier ATCC Supplier No. VR955DQ
Quantitative Genomic RNA from Human respiratory syncytial virus strain 9320 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.