Learn More
ATCC Quantitative Genomic RNA; from Influenza B virus strain; B/Wisconsin/1/2010 BX-41A
Supplier:  ATCC VR1885DQ
Encompass
Quantitative genomic RNA from Influenza B virus (Yamagata Lineage) strain B/Wisconsin/1/2010 BX-41A can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2618