Learn More
ATCC Quantitative Genomic RNA; from Human enterovirus 71; Strain: H
Catalog No. 502384062
Encompass
Quantitative Genomic RNA from Human enterovirus 71 strain H can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4062 Supplier ATCC Supplier No. VR1432DQ
Quantitative Genomic RNA from Human enterovirus 71 strain H can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.