Learn More
ATCC Quantitative Genomic RNA from; Influenza B virus; Strain: B/Hong Kong/5/72
Catalog No. 502384238
Encompass
Genomic RNA isolated from a preparation of Influenza B virus strain B/Hong Kong/5/72. This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4238 Supplier ATCC Supplier No. VR823DQ
Genomic RNA isolated from a preparation of Influenza B virus strain B/Hong Kong/5/72. This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.