Learn More
ATCC Quantitative Genomic RNA from; Human Respiratory; Syncytial Virus; Strain: Long
Supplier:  ATCC VR26DQ
Encompass
Quantitative genomic RNA from Human respiratory syncytial virus strain Long (ATCC VR-26) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4370
 Restricted Item