Learn More
ATCC Quantitative Genomic RNA from; Escherichia coli phage MS2
Catalog No. 502384382
Encompass
Quantitative Genomic RNA from Escherichia coli bacteriophage MS2 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral.
Catalog No. 50-238-4382 Supplier ATCC Supplier No. 15597B1DQ
Add to Cart
Add to Cart
Quantitative Genomic RNA from Escherichia coli bacteriophage MS2 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral.