Learn More
ATCC Quantitative Genomic; RNA from Human; coronavirus 229E
Catalog No. 502382055
Encompass
Quantitative genomic RNA from Human coronavirus 229E (ATCC VR-740) can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2055 Supplier ATCC Supplier No. VR740DQ
Quantitative genomic RNA from Human coronavirus 229E (ATCC VR-740) can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.