Learn More
ATCC Quantitative Genomic RNA; from Influenza A; virus (H3N2); strain A/Wisconsin/15/2009
Catalog No. 502382329
Encompass
Quantitative Genomic RNA from Influenza A virus (H3N2) strain A/Wisconsin/15/2009 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2329 Supplier ATCC Supplier No. VR1882DQ
Quantitative Genomic RNA from Influenza A virus (H3N2) strain A/Wisconsin/15/2009 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.