Learn More
ATCC Quantitative Genomic; RNA from Influenza; A virus (H1N1pdm); strain A/California/07;/2009 NYMC X-179A
Catalog No. 502382175
Encompass
Quantitative genomic RNA from Influenza A virus (H1N1pdm) strain A/California/07/2009 NYMC X-179A can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2175 Supplier ATCC Supplier No. VR1884DQ
Quantitative genomic RNA from Influenza A virus (H1N1pdm) strain A/California/07/2009 NYMC X-179A can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.