Learn More
ATCC Quantitative Genomic RNA from; Enterovirus 68; Strain: Fermon
Catalog No. 502381982
Encompass
Quantitative Genomic RNA from Enterovirus 68 strain Fermon can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-1982 Supplier ATCC Supplier No. VR1826DQ
Quantitative Genomic RNA from Enterovirus 68 strain Fermon can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.