Learn More
ATCC Quantitative Genomic RNA from; Human rhinovirus 2; Strain: HGP
Catalog No. 502383446
Encompass
Quantitative genomic RNA Human rhinovirus 2 strain HGP (ATCC VR-482) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3446 Supplier ATCC Supplier No. VR482DQ
Quantitative genomic RNA Human rhinovirus 2 strain HGP (ATCC VR-482) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.