Learn More
ATCC Quantitative Genomic RNA from; Human rhinovirus 1A; Strain: 2060
Catalog No. 502384718
Encompass
Quantitative Genomic RNA from Human rhinovirus 1A strain 2060 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4718 Supplier ATCC Supplier No. VR1559DQ
Quantitative Genomic RNA from Human rhinovirus 1A strain 2060 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.