Learn More
ATCC Quantitative Genomic RNA; from Influenza B virus; (Yamagata lineage); strain B/Wisconsin/1/2010
Catalog No. 502384767
Encompass
Quantitative genomic RNA from Influenza B virus (Yamagata lineage) strain B/Wisconsin/1/2010 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4767 Supplier ATCC Supplier No. VR1883DQ
Quantitative genomic RNA from Influenza B virus (Yamagata lineage) strain B/Wisconsin/1/2010 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.