Learn More
ATCC Quantitative Genomic RNA; from Betacoronavirus 1; Strain OC43
Supplier:  ATCC VR1558DQ
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2210