Learn More
ATCC Quantitative Genomic RNA from Influenza B virus strain B/Florida/4/2006 (ATCC VR-1804DQ™)
Catalog No. 502383879
Encompass
Quantitative Genomic RNA from Influenza B virus strain B/Florida/4/2006 (ATCC VR-1804DQ™) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3879 Supplier ATCC Supplier No. VR1804DQ
Quantitative Genomic RNA from Influenza B virus strain B/Florida/4/2006 (ATCC VR-1804DQ™) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.