Learn More
ATCC Quantitative Genomic RNA from Human respiratory syncytial virus strain A2 (ATCC VR-1540DQ™)
Supplier:  ATCC VR1540DQ
Encompass
Quantitative Genomic RNA from Human respiratory syncytial virus strain A2 (ATCC VR-1540DQ™) is a molecular standard used for qPCR assay development, validation, and viral load quantification. Prepared from ATCC VR-1540, this product contains ≥1 × 105 copies/μL and is suitable for respiratory disease research.
Catalog No. 50-238-2423