Learn More
ATCC Quantitative Genomic; RNA from Influenza; A virus strain; A/Florida/3/2006 (H1N1)
Catalog No. 502383128
Encompass
Quantitative genomic RNA from Influenza A virus (H1N1) strain A/Florida/3/2006 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3128 Supplier ATCC Supplier No. VR1893DQ
Quantitative genomic RNA from Influenza A virus (H1N1) strain A/Florida/3/2006 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.