Learn More
ATCC Quantitative Genomic; RNA from Human; parainfluenzavirus 3; strain: C 243
Supplier:  ATCC VR93DQ
Encompass
Quantitative Genomic RNA from Human parainfluenzavirus 3 strain C 243 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-1858