Learn More
ATCC Quantitative Genomic RNA; from Human parainfluenza; virus 1 strain C35
Catalog No. 502382392
Encompass
Quantitative Genomic RNA from Human parainfluenza virus 1 strain C35 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2392 Supplier ATCC Supplier No. VR94DQ
Quantitative Genomic RNA from Human parainfluenza virus 1 strain C35 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.