Learn More
ATCC Quantitative Genomic; RNA from Human; rhinovirus 16; Strain: 11757
Catalog No. 502381924
Encompass
Quantitative genomic RNA from Human rhinovirus 16 strain 11757 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-1924 Supplier ATCC Supplier No. VR283DQ
Quantitative genomic RNA from Human rhinovirus 16 strain 11757 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.