Learn More
ATCC Quantitative Genomic; RNA from Influenza; A virus (H1N1); Strain: A/PR/8/34
Catalog No. 502384101
Encompass
Quantitative Genomic RNA from Influenza A virus (H1N1) strain A/PR/8/34 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4101 Supplier ATCC Supplier No. VR1469DQ
Quantitative Genomic RNA from Influenza A virus (H1N1) strain A/PR/8/34 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.