Learn More
ATCC Quantitative Genomic; RNA from Influenza; A virus (H1N1); Strain: A/Virginia/ATCC1/2009
Catalog No. 502384490
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4490 Supplier ATCC Supplier No. 502384490
Add to Cart
Add to Cart
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.