Learn More
ATCC Quantitative Genomic; RNA from Influenza; B virus; Strain: B/Lee/40
Catalog No. 502381813
Encompass
Quantitative Genomic RNA from Influenza B virus strain B/Lee/40 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-1813 Supplier ATCC Supplier No. VR101DQ
Quantitative Genomic RNA from Influenza B virus strain B/Lee/40 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.