Learn More
ATCC Quantitative Genomic; DNA from; Mycoplasma hominis; Strain: PG21
Catalog No. 502382344
Encompass
Quantitative genomic DNA from Mycoplasma hominis strain PG21 (ATCC 23114). This whole-genome sequenced product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-2344 Supplier ATCC Supplier No. 23114DQ
Quantitative genomic DNA from Mycoplasma hominis strain PG21 (ATCC 23114). This whole-genome sequenced product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.