Learn More
Waters Corp Waters Corporation Standard, Quantitative Peptide Retention, Reversed Phase, UV/MS Detection, Solid
Catalog No. 502037722 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Quantitative Peptide Retention Standard is a quantitative standard designed for the calibration, development, and troubleshooting of chromatographic separations, ensuring confidence in results. It is rigorously QC tested for purity and quantitative formulation.
  • Peak retention for chromatographic reproducibility
  • UV absorptivity for signal reproducibility
  • Low to high mass range for MS
  • Water solubility
  • Tryptic-like peptides for peptide mapping studies
Catalog No. 50-203-7722 Supplier Waters Corp Supplier No. 186006555
The Quantitative Peptide Retention Standard is a quantitative standard designed for the calibration, development, and troubleshooting of chromatographic separations, ensuring confidence in results. It is rigorously QC tested for purity and quantitative formulation.
  • Peak retention for chromatographic reproducibility
  • UV absorptivity for signal reproducibility
  • Low to high mass range for MS
  • Water solubility
  • Tryptic-like peptides for peptide mapping studies