Learn More
ATCC Quantitative Synthetic; African swine fever virus DNA
Catalog No. 502384210
Encompass
Quantitative Synthetic African swine fever virus DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. Preparation includes fragments from B646L, A489, 505-2R, C717R, B962L, B119L, G1340L and D1133L genomic regions.
Catalog No. 50-238-4210 Supplier ATCC Supplier No. VR3283SD
Quantitative Synthetic African swine fever virus DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. Preparation includes fragments from B646L, A489, 505-2R, C717R, B962L, B119L, G1340L and D1133L genomic regions.