Learn More
ATCC Quantitative Synthetic; Chlamydia trachomatis; LGV Type 3 DNA
Supplier:  ATCC BAA4003SD
Encompass
Quantitative Synthetic Chlamydia trachomatis LGV Type 3 DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. Preparation includes fragments from MOMP, 16S rRNA, pmpH, and dnaB regions.
Catalog No. 50-238-4496