Learn More
ATCC Quantitative Synthetic; Chlamydia trachomatis; LGV Type 1 DNA
Catalog No. 502381777
Encompass
Quantitative Synthetic Chlamydia trachomatis LGV Type 1 DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. Preparation includes fragments from MOMP, 16S rRNA, pmpH, dnaB, putative virulence plasmid integrase regions, and conserved hypothetical virulence plasmid protein.
Catalog No. 50-238-1777 Supplier ATCC Supplier No. BAA4001SD
 Restricted Item
Add to Cart
Add to Cart
Quantitative Synthetic Chlamydia trachomatis LGV Type 1 DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. Preparation includes fragments from MOMP, 16S rRNA, pmpH, dnaB, putative virulence plasmid integrase regions, and conserved hypothetical virulence plasmid protein.