Learn More
ATCC Quantitative Synthetic; Coxiella burnetii; DNA
Catalog No. 502384207
Encompass
Quantitative Synthetic Coxiella burnetii DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. Preparation includes fragments from com1, icd, transposase (IS1111A), gyrA, and sodB regions.
Catalog No. 50-238-4207 Supplier ATCC Supplier No. BAA4000SD
Add to Cart
Add to Cart
Quantitative Synthetic Coxiella burnetii DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. Preparation includes fragments from com1, icd, transposase (IS1111A), gyrA, and sodB regions.