Learn More
ATCC Quantitative Synthetic; Dientamoeba fragilis DNA
Catalog No. 502383386
Encompass
Quantitative Synthetic DNA from Dientamoeba fragilis can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load. Preparation includes fragments from the 18S ribosomal RNA, internal transcribed spacer 1 (ITS1), and 5.8S ribosomal RNA regions.
Catalog No. 50-238-3386 Supplier ATCC Supplier No. PRA3007SD
Quantitative Synthetic DNA from Dientamoeba fragilis can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load. Preparation includes fragments from the 18S ribosomal RNA, internal transcribed spacer 1 (ITS1), and 5.8S ribosomal RNA regions.