Learn More
ATCC Quantitative Synthetic; Giardia lamblia DNA
Supplier:  ATCC PRA3006SD
Encompass
Quantitative Synthetic DNA from Giardia lamblia can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load. Preparation includes fragments from the 18S ribosomal RNA, beta-giardin, triosephosphate isomerase, and glutamate dehydrogenase regions.
Catalog No. 50-238-3910