Learn More
ATCC Quantitative Synthetic; Human coronavirus; Strain NL63 RNA
Catalog No. 502381808
Encompass
Quantitative synthetic Human coronavirus NL63 RNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from NSP3 (ORF 1A), Rdrp (nsp12), NTPase (nsp13), nsp16, spike protein, nucleocapsid, and 3’ untranslated regions.
Catalog No. 50-238-1808 Supplier ATCC Supplier No. VR3263SD
Quantitative synthetic Human coronavirus NL63 RNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from NSP3 (ORF 1A), Rdrp (nsp12), NTPase (nsp13), nsp16, spike protein, nucleocapsid, and 3’ untranslated regions.