Learn More
ATCC Quantitative Synthetic; Human herpesvirus; 6A DNA
Supplier:  ATCC VR3264SD
Encompass
Quantitative synthetic Human herpesvirus 6A DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from U31 (large tegument protein), U38 (DNA polymerase), U57 (major capsid protein), U65/U66, U67, U90 (IE1 / IE-A protein), and U94 regions.
Catalog No. 50-238-3948