Learn More
ATCC Quantitative Synthetic; Human herpesvirus; 8 DNA
Catalog No. 502381804
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. Preparation includes genomic segments taken from the minor capsid protein (ORF 26) and the latency-associated nuclear antigen (LANA or ORF 73).
Catalog No. 50-238-1804 Supplier ATCC Supplier No. VR3261SD
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. Preparation includes genomic segments taken from the minor capsid protein (ORF 26) and the latency-associated nuclear antigen (LANA or ORF 73).