Learn More
ATCC Quantitative Synthetic; Human immunodeficiency; virus 2 (HIV-2) RNA
Supplier:  ATCC VR3266SD
Encompass
Quantitative synthetic Human immunodeficiency virus 2 RNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative RT-PCR (qRT-PCR) to determine viral load. This preparation includes fragments from the envelope (ENV), group specific antigen (GAG), and DNA polymerase (POL) regions.
Catalog No. 50-238-2496