Learn More
ATCC Quantitative Synthetic Monkeypox virus DNA (ATCC VR-3270SD™)
Supplier:  ATCC VR3270SD
Encompass
Quantitative Synthetic Monkeypox virus DNA (ATCC VR-3270SD™) is a synthetic molecular standard designed for assay development, validation, and viral load quantification. This product contains fragments from the J2L, D14L, F3L, F8L, A27L, A29L, B6R, B7R, and N3R regions and it is compatible with several published qPCR protocols.
Catalog No. 50-238-2498