Learn More
ATCC Quantitative Synthetic; Mycobacterium leprae DNA
Supplier:  ATCC BAA4009SD
Encompass
Quantitative synthetic Mycobacterium leprae DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. Preparation includes fragments from the RLEP, Ag85B, 16S rRNA, and rpoB regions.
Catalog No. 50-238-1775