Learn More
ATCC Quantitative Synthetic; Parvovirus B19 DNA
Supplier:  ATCC VR3281SD
Encompass
Quantitative Synthetic Parvovirus B19 DNA, genotype 1 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the VP1, VP2, and NS1 regions and targets genotype 1.
Catalog No. 50-238-2388